3-(3,4-Dihydroxyphenyl)-L-alanine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dopar, L-dopa, L-β-3,4-Dihydroxyphenyl-Alanine, 3-Hydroxy-L-tyrosine, Levodopa

16753-0250 16753-0050 16753-1000
TS16753-0250EA 223 USD
TS16753-0250 TS16753-0050 TS16753-1000
3-(3,4-Dihydroxyphenyl)-L-alanine 99%
3-(3,4-Dihydroxyphenyl)-L-alanine
Formula: C₉H₁₁NO₄
Melting Pt: 295 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002598
CAS Number: 59-92-7
EINECS: 200-445-2

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 %
Sulfated ash ≤0.1 %
HPLC ≥98.5 %
Titration with HClO4 ≥98.5 % (On dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR