Lauric acid 99%

Supplier: Thermo Scientific Chemicals
Danger

16728-0010 16728-0050 16728-0051 16728-1000 16728-5000
TS16728-0010EA 161 USD
TS16728-0010 TS16728-0050 TS16728-0051 TS16728-1000 TS16728-5000
Lauric acid 99%
Lauric acid
Formula: C₁₂H₂₄O₂
Boiling Pt: 225 °C (100 mmHg)
Melting Pt: 44…46 °C
Flash Pt: 156 °C
Storage Temperature: Ambient
MDL Number: MFCD00002736
CAS Number: 143-07-7
EINECS: 205-582-1
Merck Index: 13,05400

Order Now

Specification Test Results

Appearance White Crystalline powder or flakes
Infrared spectrum Conforms
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR