Adonitol 98%

Supplier: Thermo Scientific Chemicals

16703-0050 16703-0100 16703-1000 16703-5000
TS16703-0050EA 39 USD
TS16703-0050 TS16703-0100 TS16703-1000 TS16703-5000
Adonitol 98%
Adonitol
Formula: HOH₂C(CH(OH))₃CH₂OH
Melting Pt: 101…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00064291
CAS Number: 488-81-3
EINECS: 207-685-7
Merck Index: 13,00168

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 101 to 105 °C
HPLC ≥97.5 % (ELSD)
Oxygen (O) 7 to 7.5 %
Conductivity 500 to 700 S/m


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR