Acetonitrile-[D₃] (99% D) for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

16623-0250 16623-0500 166230100 166232500
TS16623-0250EA 202 USD
TS16623-0250 TS16623-0500 TS166230100 TS166232500
Acetonitrile-[D₃] (99% D) for NMR spectroscopy
Acetonitrile-[D3]
Formula: D₃CCN
Boiling Pt: 79…81 °C (760 mmHg)
Melting Pt: –46 °C
Density: 0.844 g/cm³ (20 °C)
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00001881
CAS Number: 2206-26-0
EINECS: 218-616-5
UN: 1648
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.1 % (Coulometric)
D-Enrichment ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR