Thiazole 99%

Supplier: Thermo Scientific Chemicals
Warning

16613-0010 16613-0050 16613-0250
TS16613-0010EA 41 USD
TS16613-0010 TS16613-0050 TS16613-0250
Thiazole 99%
Thiazole
Formula: C₃H₃NS
Boiling Pt: 117…118 °C (760 mmHg)
Density: 1.198 g/cm³
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00005315
CAS Number: 288-47-1
EINECS: 206-021-3
UN: 1993
ADR: 3,III
Merck Index: 12,09442

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5365 to 1.5395 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR