Thiazole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
16613-0010
16613-0050
16613-0250
TS16613-0010EA
41
USD
TS16613-0010
TS16613-0050
TS16613-0250
Thiazole 99%
Thiazole
Formula:
C₃H₃NS Boiling Pt: 117…118 °C (760 mmHg) Density: 1.198 g/cm³ Flash Pt: 26 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005315 CAS Number: 288-47-1 EINECS: 206-021-3 UN: 1993 ADR: 3,III Merck Index: 12,09442 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5365 to 1.5395 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...