Tetrahydrofuran 99.6% stabilized ACS

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

16424-0050 16424-0025 164240010 164240250
TS16424-0050EA 247 USD
TS16424-0050 TS16424-0025 TS164240010 TS164240250
Tetrahydrofuran 99.6% stabilized ACS
Tetrahydrofuran
Formula: C₄H₈O
Boiling Pt: 66 °C (760 mmHg)
Melting Pt: –108.4 °C
Density: 0.88 g/cm³
Flash Pt: –21 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.03 % (Coulometric)
GC ≥99.5 %
Color scale ≤10 APHA
Stabilizer 200 to 400 ppm BHT
Residue after ignition ≤0.002 %
Acidity/alkalinity ≤0.0002 meq/g
Calcium (Ca) ≤0.5 ppm
Copper (Cu) ≤0.05 ppm
Iron (Fe) ≤0.5 ppm
Lead (Pb) ≤0.05 ppm
Peroxides ≤0.0025 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR