2-Indanone 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
beta-Hydrindone, 2-Oxoindane, 1H-Inden-2(3H)-one
16411-0100
16411-0500
TS16411-0100EA
79
USD
TS16411-0100
TS16411-0500
2-Indanone 97%
2-Indanone
Formula:
C₉H₈O Boiling Pt: 220…225 °C (1013 hPa) Melting Pt: 50…57 °C Flash Pt: 100 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003792 CAS Number: 615-13-4 EINECS: 210-410-3 |
Specification Test Results
Appearance | Light yellow to brown Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 50 to 57 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...