3-Hydroxy-p-anisaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Isovanillin, Isovanicaline, 3-Hydroxy-4-methoxybenzaldehyde, Isovanillic aldehyde
16257-0050
16257-0250
16257-1000
TS16257-0050EA
26
USD
TS16257-0050
TS16257-0250
TS16257-1000
3-Hydroxy-p-anisaldehyde 98%
3-Hydroxy-p-anisaldehyde
Formula:
C₈H₈O₃ Boiling Pt: 179 °C (15 mmHg) Melting Pt: 112…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003369 CAS Number: 621-59-0 EINECS: 210-694-9 |
Specification Test Results
Appearance | Off-white to grey to light brown Powder |
Infrared spectrum | Conforms |
Melting point | 112 to 116 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...