3-Hydroxy-p-anisaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Isovanillin, Isovanicaline, 3-Hydroxy-4-methoxybenzaldehyde, Isovanillic aldehyde

16257-0050 16257-0250 16257-1000
TS16257-0050EA 26 USD
TS16257-0050 TS16257-0250 TS16257-1000
3-Hydroxy-p-anisaldehyde 98%
3-Hydroxy-p-anisaldehyde
Formula: C₈H₈O₃
Boiling Pt: 179 °C (15 mmHg)
Melting Pt: 112…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00003369
CAS Number: 621-59-0
EINECS: 210-694-9

Order Now

Specification Test Results

Appearance Off-white to grey to light brown Powder
Infrared spectrum Conforms
Melting point 112 to 116 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR