Diisopropyl ether 98+% stabilised, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Isopropyl ether

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

15896-0010 158960025 158960100
TS15896-0010EA 91 USD
TS15896-0010 TS158960025 TS158960100
Diisopropyl ether 98+% stabilised, pure
Diisopropyl ether
Formula: (CH₃)₂CHOCH(CH₃)₂
MW: 102.18 g/mol
Boiling Pt: 68.5
Melting Pt: -86
Density: 0.72 g/cm³
Flash Pt: –29 °C
Storage Temperature: Ambient
MDL Number: MFCD00008880
CAS Number: 108-20-3
EINECS: 203-560-6
UN: 1159
ADR: 3 II
Merck Index: 13,05232
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=932b8c021b2bfa9a11b8d8812cafbafecc6f9239&vid=dea267e876203eb2c2674787701ec602da1b3cf6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3670 to 1.3690 (20°C, 589 nm)
Water ≤0.5 %
GC ≥98.0 %
Color scale ≤15 APHA
Stabilizer 5 to 150 ppm BHT
Acidity ≤0.005 %
Peroxides ≤0.002 % (as H2O2)
Mercury (Hg) ≤0.0001 %
Zinc (Zn) ≤0.001 %
Heavy metals (as Pb) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR