2-Hydroxyhippuric acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(2-Hydroxybenzoyl)glycine

15862-0025 15862-0100
TS15862-0025EA 95 USD
TS15862-0025 TS15862-0100
2-Hydroxyhippuric acid 95%
2-Hydroxyhippuric acid
Formula: C₉H₉NO₄
MW: 195.17 g/mol
Melting Pt: 164…169 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002695
CAS Number: 487-54-7
EINECS: 207-661-6

Order Now

Specification Test Results

Appearance White to cream Powder
Infrared spectrum Conforms
Melting point 163 to 168 °C
HPLC ≥94 %
Calcium (Ca) ≤0.02 %
Iron (Fe) ≤0.005 %
Magnesium (Mg) ≤0.02 %
Potassium (K) ≤0.02 %
Heavy metals (as Pb) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR