Pyridinium tribromide 90+%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Pyridinium bromide perbromide, Monopyridin-1-ium tribromide, Pyridine hydrobromide perbromide
15836-5000
15836-1000
TS15836-5000EA
345
USD
TS15836-5000
TS15836-1000
Pyridinium tribromide 90+%, Technical Grade
Pyridinium tribromide
Formula:
C₅H₆Br₃N MW: 319.82 g/mol Melting Pt: 127…133 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013223 CAS Number: 39416-48-3 EINECS: 254-446-8 UN: 3261 ADR: 8 II |
Specification Test Results
Appearance | Orange to red to brown Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 130 to 137 °C (Dec) |
Titration Iodometric | ≥90.0 % |
UV | (0.1 M in water) |
pH | 3.8 to 4.5 (1 M in water) |
Titration with NaOH | ≥99.4 % |
Chloride (Cl) | ≤0.005 % |
Iron (Fe) | ≤0.0005 % |
Heavy metals (as Pb) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...