Isobutanol 99+%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

158280010 158280025 158280250
TS158280010EA 78 USD
TS158280010 TS158280025 TS158280250
Isobutanol 99+%, Extra Pure
Isobutanol
Formula: (CH₃)₂CHCH₂OH
Boiling Pt: 108 °C (760 mmHg)
Melting Pt: –108 °C
Density: 0.8 g/cm³
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00004740
CAS Number: 78-83-1
EINECS: 201-148-0
UN: 1212
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3945 to 1.3975 (20°C, 589 nm)
Water ≤0.2 % (K.F.)
Density (20°C) 0.800 to 0.804 g/ml
GC ≥99.0 %
Color scale ≤10 APHA
Residue after evaporation ≤0.005 %
Insoluble matter ≤0.01 %
Residue after reduction ≤0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR