Pyrrole 99%, extra pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1H-Pyrrole
15771-1000
15771-5000
15771-0025
15771-0250
TS15771-1000EA
94
USD
TS15771-1000
TS15771-5000
TS15771-0025
TS15771-0250
Pyrrole 99%, extra pure
Pyrrole
Formula:
C₄H₅N MW: 67.09 g/mol Boiling Pt: 129…131 °C (760 mmHg) Melting Pt: -24 Density: 0.96 g/cm³ Flash Pt: 36 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005216 CAS Number: 109-97-7 EINECS: 203-724-7 UN: 1992 ADR: 3 III Merck Index: 12,08198 |
Specification Test Results
Appearance | Clear colorless to brown Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5080 to 1.5100 (20°C, 589 nm) |
Water | ≤0.15 % |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...