Pyrrole 99%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1H-Pyrrole

15771-1000 15771-5000 15771-0025 15771-0250
TS15771-1000EA 94 USD
TS15771-1000 TS15771-5000 TS15771-0025 TS15771-0250
Pyrrole 99%, extra pure
Pyrrole
Formula: C₄H₅N
MW: 67.09 g/mol
Boiling Pt: 129…131 °C (760 mmHg)
Melting Pt: -24
Density: 0.96 g/cm³
Flash Pt: 36 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005216
CAS Number: 109-97-7
EINECS: 203-724-7
UN: 1992
ADR: 3 III
Merck Index: 12,08198

Order Now

Specification Test Results

Appearance Clear colorless to brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5080 to 1.5100 (20°C, 589 nm)
Water ≤0.15 %
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR