1,10-Phenanthroline 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
o-Phenanthroline
15753-0250
15753-0050
15753-1000
15753-5000
TS15753-0250EA
128
USD
TS15753-0250
TS15753-0050
TS15753-1000
TS15753-5000
1,10-Phenanthroline 99+%
1,10-Phenanthroline
Formula:
C₁₂H₈N₂ Melting Pt: 117…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011678 CAS Number: 66-71-7 EINECS: 200-629-2 UN: 2811 ADR: 6.1,III Merck Index: 12,07356 |
|
Specification Test Results
Appearance | White to light yellow to light pink Powder |
Infrared spectrum | Conforms |
Melting point | 114 to 120 °C |
Sulfated ash | ≤0.1 % |
Titration with HClO4 | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...