Methylcyclopentadiene dimer 95%

Supplier: Thermo Scientific Chemicals
Warning

15685-0010 15685-0025 15685-2500
TS15685-0010EA 226 USD
TS15685-0010 TS15685-0025 TS15685-2500
Methylcyclopentadiene dimer 95%
Methylcyclopentadiene dimer
Formula: C₁₂H₁₆
Boiling Pt: 200 °C (760 mmHg)
Melting Pt: –51 °C
Density: 0.94 g/cm³
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00082297
CAS Number: 26472-00-4
EINECS: 247-724-5
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
NMR (1H-NMR) ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR