2-Ethoxyethanol 99%, Extra Pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
15602-0010
156020025
156020250
TS15602-0010EA
76
USD
TS15602-0010
TS156020025
TS156020250
2-Ethoxyethanol 99%, Extra Pure
2-Ethoxyethanol
Formula:
C₂H₅OCH₂CH₂OH Boiling Pt: 135 °C (760 mmHg) Melting Pt: –70 °C Density: 0.93 g/cm³ Flash Pt: 40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002869 CAS Number: 110-80-5 EINECS: 203-804-1 UN: 1171 ADR: 3,III REACH: 01-2119560582-38 Merck Index: 13,03786 |
|
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4060 to 1.4100 (20°C, 589 nm) |
Water | ≤0.15 % (K.F.) |
GC | ≥98.5 % |
Color scale | ≤10 APHA |
Specific gravity | (20°C) 0.928 to 0.932 |
Alkalinity | ≤0.0002 meq/g |
Substance darkened by H2SO4 | passes test |
Calcium (Ca) | ≤0.5 ppm |
Chromium (Cr) | ≤0.02 ppm |
Cobalt (Co) | ≤0.02 ppm |
Copper (Cu) | ≤0.05 ppm |
Iron (Fe) | ≤0.2 ppm |
Magnesium (Mg) | ≤0.1 ppm |
Manganese (Mn) | ≤0.02 ppm |
Nickel (Ni) | ≤0.02 ppm |
Zinc (Zn) | ≤0.1 ppm |
Heavy metals (as Pb) | ≤1 ppm |
Ethanol | 99.8 to 100 % v/v (calculated) |
Substance reducing dichromate | passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...