4-Methylbenzyl alcohol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Hydroxymethyl)toluole, p-Tolylmethanol, (4-Methylphenyl)methanol

15592-0050 15592-0250 15592-1000
TS15592-0050EA 20 USD
TS15592-0050 TS15592-0250 TS15592-1000
4-Methylbenzyl alcohol 98%
4-Methylbenzyl alcohol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 217
Melting Pt: 59
Storage Temperature: Ambient
MDL Number: MFCD00004664
CAS Number: 589-18-4
EINECS: 209-639-1

Order Now

Specification Test Results

Appearance White Crystals or needles or crystalline mass
Infrared spectrum Conforms
Melting point 56 to 61 °C
GC ≥97.5 %
UV at 400 nm A: ≤0.01
Color scale ≤15 APHA
Residue after evaporation ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR