1-Naphthylmethylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Naphthalenemethylamine, Naphthalen-1-ylmethanamine, Naphthalene-1-methylamine, 1-(Aminomethyl)naphthalene

15558-0010 15558-0050
TS15558-0010EA 31 USD
TS15558-0010 TS15558-0050
1-Naphthylmethylamine 97%
1-Naphthylmethylamine
Formula: C₁₁H₁₁N
MW: 157.22 g/mol
Boiling Pt: 92
Melting Pt: 267…269 °C
Density: 1.073 g/cm³
Flash Pt: >110 °C
MDL Number: MFCD00004048
CAS Number: 118-31-0
EINECS: 204-244-0
UN: 2735

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6420 to 1.6440 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR