Cyclohexene 99% stabilised, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
15484-0010
154840100
154845000
TS15484-0010EA
111
USD
TS15484-0010
TS154840100
TS154845000
Cyclohexene 99% stabilised, pure
Cyclohexene
Formula:
C₆H₁₀ MW: 82.15 g/mol Boiling Pt: 83 Melting Pt: -104 Density: 0.81 g/cm³ Flash Pt: –20 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001539 CAS Number: 110-83-8 EINECS: 203-807-8 UN: 2256 ADR: 3 II Merck Index: 13,02756 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4450 to 1.4480 (20°C, 589 nm) |
GC | ≥98.5 % |
Stabilizer | Approx. 100 ppm 2-tert-Butyl-4-methylphenol |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...