2,2'-Butyliminodiethanol 98%

Supplier: Thermo Scientific Chemicals
Danger

15436-0025 15436-2500 15436-0010
TS15436-0025EA 196 USD
TS15436-0025 TS15436-2500 TS15436-0010
2,2'-Butyliminodiethanol 98%
2,2'-Butyliminodiethanol
Formula: H₃CCH₂CH₂CH₂N(CH₂CH₂OH)₂
Boiling Pt: 264…278 °C (760 mmHg)
Melting Pt: –70 °C
Density: 0.968 g/cm³
Flash Pt: 143 °C
Storage Temperature: Ambient
MDL Number: MFCD00002856
CAS Number: 102-79-4
EINECS: 203-055-0
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4610 to 1.4640 (20°C, 589 nm)
Water ≤0.5 %
GC ≥97.5 %
Color scale ≤200 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR