1-Adamantanecarbonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

15248-0050 15248-0250
TS15248-0050EA 48 USD
TS15248-0050 TS15248-0250
1-Adamantanecarbonyl chloride 97%
1-Adamantanecarbonyl chloride
Formula: C₁₁H₁₅ClO
Boiling Pt: 135…136 °C (10 mmHg)
Melting Pt: 48…52 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00074724
CAS Number: 2094-72-6
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance White to off-white Crystalline solid
Infrared spectrum Conforms
Melting point 48 to 52 °C
Titration RCOCl ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR