(R)-(-)-2-Aminobutyric acid 99+%

Synonyms: (R)-()-2-Aminobutyric acid, D-2-Aminobutyric acid, D(-)-2-Aminobutyric acid, H-D-Abu-OH

15214-0250
TS15214-0050EA 150.38 USD
TS15214-0050 TS15214-0250
(R)-(-)-2-Aminobutyric acid 99+%
(R)-(-)-2-Aminobutyric acid
Formula: C₄H₉NO₂
MW: 103.12 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00064414
CAS Number: 2623-91-8
EINECS: 220-084-4

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Specific optical rotation -20.2° to -22.2° (20 °C, 589 nm) c=2, 6N HCl
Titration with HClO4 ≥99.0 %
Peroxides ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR