Myricetin 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 3,3',4',5,5',7-Hexahydroxyflavone, 3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one

15159-0250
TS15159-0250EA 104 USD
TS15159-0250
Myricetin 95%
Myricetin
Formula: C₁₅H₁₀O₈
MW: 318.24 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006827
CAS Number: 529-44-2

Order Now

Specification Test Results

Appearance Brown-yellow to green-brown Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥94 %
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR