Ethylene glycol monomethyl ether (2-methoxyethanol) 99.5+% for analysis
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethylene glycol monomethyl ether
149360010
149360025
TS149360010EA
76
USD
TS149360010
TS149360025
Ethylene glycol monomethyl ether (2-methoxyethanol) 99.5+% for analysis
Ethylene glycol monomethyl ether (2-Methoxyethanol)
Formula:
C₃H₈O₂ Boiling Pt: 124 °C (760 mmHg) Melting Pt: –85 °C Density: 0.96 g/cm³ Flash Pt: 38 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002867 CAS Number: 109-86-4 EINECS: 203-713-7 UN: 1188 ADR: 3,III |
Specification Test Results
Appearance | Clear liquid |
Water | ≤0.1 % (Coulometric) |
GC | ≥99.5 % |
Color scale | ≤10 APHA |
Non-volatile matter | ≤0.002 % |
Acidity | ≤0.001 meq/g |
Alkalinity | ≤0.0005 meq/g |
Substance colored by H2SO4 | Passes test |
Aluminium (Al) | ≤5 ppm |
Barium (Ba) | ≤1 ppm |
Cadmium (Cd) | ≤0.05 ppm |
Calcium (Ca) | ≤0.5 ppm |
Chromium (Cr) | ≤0.02 ppm |
Copper (Cu) | ≤0.02 ppm |
Iron (Fe) | ≤0.5 ppm |
Lead (Pb) | ≤0.1 ppm |
Magnesium (Mg) | ≤0.1 ppm |
Manganese (Mn) | ≤0.02 ppm |
Nickel (Ni) | ≤0.02 ppm |
Zinc (Zn) | ≤0.1 ppm |
Aldehyde (C2H4O) | ≤0.003 % |
Peroxides | ≤20 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...