Amyl acetate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: n-Pentyl acetate, Acetic acid amyl ester, Acetic acid pentyl ester, Pentyl acetate

14918-0010 149180025 149182500
TS14918-0010EA 205 USD
TS14918-0010 TS149180025 TS149182500
Amyl acetate 99%
Amyl acetate
Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 149
Melting Pt: -71
Density: 0.87 g/cm³
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00009500
CAS Number: 628-63-7
EINECS: 211-047-3
UN: 1104
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37af9fce9b4d1452153c5c88d6e47d0d5e08f221&vid=b0e4dd5050dab83dda2db704a160c2058b4bd15a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4010 to 1.4030 (20°C, 589 nm)
GC ≥98.5 %
TLC ≥97.5 %
Specific optical rotation -10.8° to -11.8° (20°C, 589 nm) (c=2 in DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR