Benzoic acid 99%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

14913-0010 14913-0025 14913-0250 14913-5000
TS14913-0010EA 73 USD
TS14913-0010 TS14913-0025 TS14913-0250 TS14913-5000
Benzoic acid 99%, extra pure
Benzoic acid
Formula: C₇H₆O₂
Boiling Pt: 249 °C (760 mmHg)
Melting Pt: 121…123 °C
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00002398
CAS Number: 65-85-0
EINECS: 200-618-2
REACH: 01-2119455536-33
Merck Index: 13,01092
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d561f311b11ea8ab2c8f07253e6748e588eb71aa&vid=dcf5e45072cda5106216b497cd29716270ae772c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to off-white Flakes
Infrared spectrum Conforms
Melting point 121 to 124 °C
Water ≤0.5 % (K.F.)
Sulfated ash ≤0.01 %
Titration with NaOH ≥98.5 %
Clarity of alcohol solution (20 wt.% soln. in ethanol) clear
Color of molten substance ≤50 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR