Triiodomethane 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Iodoform
AC14912-0025
AC14912-1000
AC14912-5000
TS14912-0025EA
961
USD
TS14912-0025
TS14912-1000
TS14912-5000
Triiodomethane 99+%
Triiodomethane
Formula:
CHI₃ Melting Pt: 119…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001069 CAS Number: 75-47-8 EINECS: 200-874-5 Merck Index: 13,05055 |
Specification Test Results
Appearance | Bright yellow to dark yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 119 to 122 °C |
Loss on drying | ≤0.5 % (24 h) (Over silicagel) |
Residue after ignition | ≤0.1 % |
Titration | ≥99.0 % |
Chloride (Cl) | ≤110 ppm |
Sulfate (SO4) | ≤170 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...