Dimethyl sulfite 99%

Supplier: Thermo Scientific Chemicals
Warning

14875-1000
TS14875-1000EA 191 USD
TS14875-1000
Dimethyl sulfite 99%
Dimethyl sulphite
Formula: C₂H₆O₃S
Boiling Pt: 126 °C (1013 hPa)
Density: 1.29 g/cm³
Flash Pt: 30 °C
MDL Number: MFCD00008415
CAS Number: 616-42-2
EINECS: 210-481-0
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4085 to 1.4105 (20 °C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR