Nitromethane 99%, pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
148510025
148510100
TS14851-0010EA
181
USD
TS14851-0010
TS148510010
TS148510025
TS148510100
Nitromethane 99%, pure
Nitromethane
Formula:
CH₃NO₂ Boiling Pt: 100…102 °C (760 mmHg) Melting Pt: –29 °C Density: 1.12 g/cm³ Flash Pt: 35 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007400 CAS Number: 75-52-5 EINECS: 200-876-6 UN: 1261 ADR: 3,II Merck Index: 13,06644 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3810 to 1.3830 (20°C, 589 nm) |
GC | ≥98.5 % |
Color scale | ≤20 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...