α,α,α-Trifluoroacetophenone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,2,2-Trifluoroacetophenone, Phenacyl trifluoride, Trifluoroacetylbenzene, 1-Phenyl-2,2,2-trifluoro-1-ethanone
14835-0050
14835-0250
14835-1000
TS14835-0050EA
46
USD
TS14835-0050
TS14835-0250
TS14835-1000
α,α,α-Trifluoroacetophenone 99%
α,α,α-Trifluoroacetophenone
Formula:
C₈H₅F₃O MW: 174.12 g/mol Boiling Pt: 165 Melting Pt: -40 Density: 1.24 g/cm³ Flash Pt: 41 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000420 CAS Number: 434-45-7 EINECS: 207-103-1 UN: 1993 ADR: 3 III |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4580 to 1.4600 (20 °C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...