1-Dodecansulfonic acid sodium salt 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: Dodecane-1-sulfonic acid sodium salt, Sodium 1-dodecanesulphonate, Dodecane-1-sulphonic acid sodium salt, Sodium 1-dodecansulphonate, 1-Dodecanesulphonic acid, sodium salt, 1-Dodecanesulfonic acid, sodium salt, Sodium dodecane-1-sulphonate, Sodium dodecane-1-sulfonate, Sodium 1-dodecansulfonate, Sodium 1-dodecanesulfonate

AC14790-0010
TS14790-0010EA 81 USD
TS14790-0010
1-Dodecansulfonic acid sodium salt 99+%
1-Dodecansulphonic acid sodium salt
Formula: C₁₂H₂₅NaO₃S
MW: 272.38 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007527
CAS Number: 2386-53-0
EINECS: 219-200-6

Order Now

Specification Test Results

Appearance White to cream Powder
Infrared spectrum Conforms
Assay ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR