EDTA trisodium salt hydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Trisodium hydrogen ethylenediaminetetraacetate hydrate, Ethylenediaminetetra-acetic acid trisodium salt hydrate, Edetic acid trisodium salt hydrate, Ethylenedinitrilotetra-acetic acid trisodium salt hydrate

14786-5000
TS14786-5000EA 95 USD
TS14786-5000
EDTA trisodium salt hydrate 98%
EDTA trisodium salt hydrate
Formula: C₁₀H₁₃N₂Na₃O₈·H₂O
MW: 376.21 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149675
CAS Number: 85715-60-2
EINECS: 205-758-8

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration Complexometric ≥97.5 % (corrected for water)
Water ≤10 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR