EDTA disodium salt dihydrate 99+% for analysis

Supplier: Thermo Scientific Chemicals
Warning

14785-0010 14785-0050 14785-1000 14785-5000
TS14785-0010EA 224 USD
TS14785-0010 TS14785-0050 TS14785-1000 TS14785-5000
EDTA disodium salt dihydrate 99+% for analysis
EDTA disodium salt dihydrate
Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
Melting Pt: 252 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 205-358-3
Merck Index: 1303543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Assay 99.0 to 101.0 % (On dry substance)
Loss on drying ≤11.4 % (200°C, 3 h)
pH 4.0 to 6.0 (1 % at 20°C)
Iron (Fe) ≤80 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR