Phenolphthalein 98.5%, pure indicator grade

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

14771-0025 14771-1000 14771-5000
TS14771-0025EA 574 USD
TS14771-0025 TS14771-1000 TS14771-5000
Phenolphthalein 98.5%, pure indicator grade
Phenolphthalein
Formula: C₂₀H₁₄O₄
Melting Pt: 258…263 °C
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
EINECS: 201-004-7
REACH: 01-2119498295-24
Merck Index: 14,07243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to almost white Powder
Infrared spectrum Conforms
Assay 98.0 to 102.0 %
Loss on drying ≤0.5 % (105°C)
Sulfated ash ≤0.1 %
Acidity/alkalinity (per Ph. Eur.) Passes test
Color of solution (per Ph. Eur.) Passes test
Chloride (Cl) ≤100 ppm
Sulfate (SO4) ≤200 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR