Chloroacetyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

14729-0010 14729-0025 14729-1000 14729-2500
TS14729-0010EA 120 USD
TS14729-0010 TS14729-0025 TS14729-1000 TS14729-2500
Chloroacetyl chloride 98%
Chloroacetyl chloride
Formula: C₂H₂Cl₂O
MW: 112.94 g/mol
Boiling Pt: 105
Melting Pt: -22.5
Density: 1.42 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000725
CAS Number: 79-04-9
EINECS: 201-171-6
UN: 1752
ADR: 6.1
Merck Index: 13,02074

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4520 to 1.4540 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR