Chloroacetyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
14729-0010
14729-0025
14729-1000
14729-2500
TS14729-0010EA
120
USD
TS14729-0010
TS14729-0025
TS14729-1000
TS14729-2500
Chloroacetyl chloride 98%
Chloroacetyl chloride
Formula:
C₂H₂Cl₂O MW: 112.94 g/mol Boiling Pt: 105 Melting Pt: -22.5 Density: 1.42 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00000725 CAS Number: 79-04-9 EINECS: 201-171-6 UN: 1752 ADR: 6.1 Merck Index: 13,02074 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4520 to 1.4540 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...