2,5-Xylidine 99%

Danger

Synonyms: 2-Amino-p-xylene, 2,5-Dimethylaniline

14669-0050
TS14669-0050EA 31 USD
TS14669-0050 TS14669-1000 TS14669-5000
2,5-Xylidine 99%
2,5-Xylidine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 218
Melting Pt: 15
Density: 0.97 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00007743
CAS Number: 95-78-3
EINECS: 202-451-0
UN: 1711
ADR: 6.1 II
Merck Index: 12,10217

Order Now

Specification Test Results

Appearance Clear yellow to red-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5582 to 1.5602 (20°C, 589 nm)
GC ≥98.5 %
Titration with HClO4 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR