Safranine O 95%, pure, high purity biological stain

Supplier: Thermo Scientific Chemicals
Danger

14664-0250 14664-1000
TS14664-0250EA 69 USD
TS14664-0250 TS14664-1000
Safranine O 95%, pure, high purity biological stain
Safranine O
Formula: C₂₀H₁₉ClN₄
Storage Temperature: Ambient
MDL Number: MFCD00011759
CAS Number: 477-73-6
EINECS: 207-518-8

Order Now

Specification Test Results

Appearance Green to red-brown Powder
Infrared spectrum Conforms
Loss on drying ≤10 %
Dye content ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR