Pentafluorobenzyl bromide 97%

Supplier: Thermo Scientific Chemicals
Danger

14633-0050 14633-0100 14633-0500
TS14633-0050EA 49 USD
TS14633-0050 TS14633-0100 TS14633-0500
Pentafluorobenzyl bromide 97%
Pentafluorobenzyl bromide
Formula: (C₆F₅)CH₂Br
Boiling Pt: 174…175 °C (760 mmHg)
Melting Pt: 19 °C
Density: 1.72 g/cm³
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00000299
CAS Number: 1765-40-8
EINECS: 217-182-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4700 to 1.4720 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR