Pentafluorobenzyl bromide 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
14633-0050
14633-0100
14633-0500
TS14633-0050EA
49
USD
TS14633-0050
TS14633-0100
TS14633-0500
Pentafluorobenzyl bromide 97%
Pentafluorobenzyl bromide
Formula:
(C₆F₅)CH₂Br Boiling Pt: 174…175 °C (760 mmHg) Melting Pt: 19 °C Density: 1.72 g/cm³ Flash Pt: 82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000299 CAS Number: 1765-40-8 EINECS: 217-182-4 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4700 to 1.4720 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...