1-(2-Pyridylazo)-2-naphthol (PAN) 98%, pure indicator grade

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: PAN, 1-(2-Pyridylazo)-2-naphthol

14631-0050 14631-0100 14631-0500
TS14631-0050EA 148 USD
TS14631-0050 TS14631-0100 TS14631-0500
1-(2-Pyridylazo)-2-naphthol (PAN) 98%, pure indicator grade
1-(2-Pyridylazo)-2-naphthol
Formula: C₁₅H₁₁N₃O
Melting Pt: 138…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00004071
CAS Number: 85-85-8
EINECS: 201-637-9

Order Now

Specification Test Results

Appearance Orange to red-brown to brown Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %
Suitability (as complexom. indicator) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR