α-Tropolone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Tropolone, 2-Hydroxytropone, 2-Hydroxycyclohepta-2,4,6-trien-1-one, 1,2-Tropolone, 2-Hydroxycyclohepta-2,4,6-trienone, α-Tropolone

14058-0010 14058-0050 14058-0250
TS14058-0010EA 51 USD
TS14058-0010 TS14058-0050 TS14058-0250
α-Tropolone 98%
α-Tropolone
Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 115 °C (12 mmHg)
Melting Pt: 50…54 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004158
CAS Number: 533-75-5
EINECS: 208-577-2

Order Now

Specification Test Results

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
Melting point 50 to 54 °C (on dry substance)
GC ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR