Triethanolamine (trolamine) 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Tris-(2-hydroxyethyl)amine, 2,2',2''-Nitrilotriethanol
13956-0010
13956-0025
TS13956-0010EA
94
USD
TS13956-0010
TS13956-0025
Triethanolamine (trolamine) 97%
Triethanolamine (Trolamine)
Formula:
C₆H₁₅NO₃ Boiling Pt: 360 °C (760 mmHg) Melting Pt: 21 °C Density: 1.1245 g/cm³ Flash Pt: 190 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002855 CAS Number: 102-71-6 EINECS: 203-049-8 Merck Index: 13,09739 |
Specification Test Results
Appearance | Clear viscous liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4830 to 1.4860 (20°C, 589 nm) |
GC | ≥96.0 % |
Color scale | ≤50 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...