Triethanolamine (trolamine) 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Tris-(2-hydroxyethyl)amine, 2,2',2''-Nitrilotriethanol

13956-0010 13956-0025
TS13956-0010EA 94 USD
TS13956-0010 TS13956-0025
Triethanolamine (trolamine) 97%
Triethanolamine (Trolamine)
Formula: C₆H₁₅NO₃
Boiling Pt: 360 °C (760 mmHg)
Melting Pt: 21 °C
Density: 1.1245 g/cm³
Flash Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00002855
CAS Number: 102-71-6
EINECS: 203-049-8
Merck Index: 13,09739

Order Now

Specification Test Results

Appearance Clear viscous liquid
Infrared spectrum Conforms
Refractive index 1.4830 to 1.4860 (20°C, 589 nm)
GC ≥96.0 %
Color scale ≤50 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR