p-Toluenesulfonic acid monohydrate 99%, extra pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Methylbenzenesulphonic acid hydrate, Toluene-4-sulphonic acid monohydrate, 4-Methylbenzenesulfonic acid hydrate, Toluene-4-sulfonic acid monohydrate
13902-0025
13902-0050
13902-0100
13902-1000
13902-5000
TS13902-0025EA
130
USD
TS13902-0025
TS13902-0050
TS13902-0100
TS13902-1000
TS13902-5000
p-Toluenesulfonic acid monohydrate 99%, extra pure
p-Toluenesulphonic acid monohydrate
Formula:
C₇H₈O₃S·1H₂O Melting Pt: 100…108 °C Flash Pt: 180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00142137 CAS Number: 6192-52-5 EINECS: 203-180-0 UN: 2585 ADR: 8,III Merck Index: 12,09671 |
Specification Test Results
Appearance | White to off-white Adhering crystals (May discolor to pink on storage) |
Infrared spectrum | Conforms |
Melting point | 100 to 108 °C |
Titration Acid | The para-isomer in TSA is min. 97% |
Titration with CHA | ≤0.5 % Sulfuric acid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...