p-Toluenesulfonic acid monohydrate 99%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Methylbenzenesulphonic acid hydrate, Toluene-4-sulphonic acid monohydrate, 4-Methylbenzenesulfonic acid hydrate, Toluene-4-sulfonic acid monohydrate

13902-0025 13902-0050 13902-0100 13902-1000 13902-5000
TS13902-0025EA 130 USD
TS13902-0025 TS13902-0050 TS13902-0100 TS13902-1000 TS13902-5000
p-Toluenesulfonic acid monohydrate 99%, extra pure
p-Toluenesulphonic acid monohydrate
Formula: C₇H₈O₃S·1H₂O
Melting Pt: 100…108 °C
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00142137
CAS Number: 6192-52-5
EINECS: 203-180-0
UN: 2585
ADR: 8,III
Merck Index: 12,09671

Order Now

Specification Test Results

Appearance White to off-white Adhering crystals (May discolor to pink on storage)
Infrared spectrum Conforms
Melting point 100 to 108 °C
Titration Acid The para-isomer in TSA is min. 97%
Titration with CHA ≤0.5 % Sulfuric acid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR