Thiourea 99%, extra pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Sulfourea, Thiocarbamide, Sulphourea
13891-0010
13891-0050
13891-5000
TS13891-0010EA
88
USD
TS13891-0010
TS13891-0050
TS13891-5000
Thiourea 99%, extra pure
Thiourea
Formula:
NH₂CSNH₂ MW: 76.12 g/mol Melting Pt: 171…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008067 CAS Number: 62-56-6 EINECS: 200-543-5 UN: 3077 ADR: 9 III Merck Index: 13,09443 |
Specification Test Results
Appearance | White to off-white Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 171 to 177 °C |
Titration Iodimetric | ≥98.5 % |
Loss on drying | ≤0.5 % |
Sulfated ash | ≤0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...